Assay Details
| Gene Symbol: 8030423F21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045738.1 | 3 | 1577 | 106 | ||
| GenBank mRNA | AK033101.1 | - | 3 | 1577 | 106 | |
| AK078739.1 | - | 1 | 1249 | 106 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
leucine-rich repeat LGI family, member 2 |
Lgi2 | mKIAA1916 | -42.2kb | Sense | Chr. 5 - 52533517 - 52566462 on NCBI Build GRCm38 |
| Downstream | Sep (O-phosphoserine) tRNA:Sec (selenocysteine) tRNA synthase | Sepsecs |
9130208G10, AA986712, D5Ertd135e, SLA, SecS |
33.3kb | Sense | Chr. 5 - 52643407 - 52669770 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

8030423F21Rik
Genomic Map