Assay Details
| Gene Symbol: Dtymk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK080078.1 | - | 1 - 2 | 35 | 74 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
autophagy related 4B, cysteine peptidase |
Atg4b |
2510009N07Rik, AW048066, Apg4b, Atg4bl, Autl1 |
-8.9kb | Sense | Chr. 1 - 93755033 - 93789529 on NCBI Build GRCm38 |
| Downstream |
inhibitor of growth family, member 5 |
Ing5 |
1700001C14Rik, 1700027H23Rik, 1810018M11Rik, AI225768 |
2.7kb | Sense | Chr. 1 - 93802051 - 93822101 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Dtymk
Genomic Map