Assay Details
| Gene Symbol: Mob3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK080169.1 | - | 1 - 2 | 34 | 170 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | MAP kinase-interacting serine/threonine kinase 2 | Mknk2 |
2010016G11Rik, Gprk7, Mnk2 |
-23.5kb | Sense | Chr. 10 - 80665318 - 80676293 on NCBI Build GRCm38 |
| Downstream | IZUMO family member 4 | Izumo4 |
9030607L17Rik, AI550407 |
371.0 | Anti-sense | Chr. 10 - 80702199 - 80705382 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Mob3a
Genomic Map