Assay Details
| Gene Symbol: Sppl2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023220.2 | NP_075709.2 | 15 | 2980 | 101 | |
| GenBank mRNA | AK080421.1 | - | 1 | 1050 | 101 | |
| AK081953.1 | - | 1 | 1092 | 101 | ||
| AK152512.1 | - | 1 | 192 | 101 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
transient receptor potential cation channel, subfamily M, member 7 |
Trpm7 |
2310022G15Rik, 4833414K03Rik, 5033407O22Rik, CHAK, CHAK1, LTrpC-7, Ltpr7, Ltrpc7, TRPPLIK |
-16.7kb | Sense | Chr. 2 - 126791558 - 126876261 on NCBI Build GRCm38 |
| Downstream | microRNA 3473g | Mir3473g |
Gm27751, mmu-mir-3473g |
8.2kb | Sense | Chr. 2 - 126902250 - 126902385 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Sppl2a
Genomic Map