Assay Details
| Gene Symbol: A730063M14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK080508.1 | - | 2 - 3 | 1308 | 59 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | tetraspanin 31 | Tspan31 |
2700085A14Rik, AW742554, Sas |
-1.2kb | Anti-sense | Chr. 10 - 127067290 - 127070261 on NCBI Build GRCm38 |
| Downstream |
ArfGAP with GTPase domain, ankyrin repeat and PH domain 2 |
Agap2 |
AGAP-2, Centg1, PIKE, cnt-g1, mKIAA0167 |
462.0 | Sense | Chr. 10 - 127075258 - 127093173 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

A730063M14Rik
Genomic Map