Assay Details
| Gene Symbol: A830010M09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK080594.1 | - | 1 | 849 | 108 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | N-acetylated alpha-linked acidic dipeptidase-like 2 | Naaladl2 |
2810043G22Rik, EG635702, Gm1021 |
-1.1mb | Sense | Chr. 3 - 23798103 - 24784006 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA A830092H15 gene | A830092H15Rik | 464.4kb | Anti-sense | Chr. 3 - 26331953 - 26336870 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

A830010M09Rik
Genomic Map