Assay Details
| Gene Symbol: B930093H17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK081132.1 | - | 1 | 655 | 77 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 28b | Mir28b | mmu-mir-28b | -71.4kb | Anti-sense | Chr. 8 - 73016512 - 73016591 on NCBI Build GRCm38 |
| Downstream | uncharacterized LOC102640619 | LOC102640619 | 1.5mb | Sense | Chr. 8 - 74590987 - 74833947 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

B930093H17Rik
Genomic Map