Assay Details
| Gene Symbol: Dcaf12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK083369.1 | - | 1 | 1347 | 62 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ubiquitin-associated protein 2 | Ubap2 |
1190005K07Rik, AA408600, AU045235, UBAP-2, mKIAA1491 |
-36.3kb | Sense | Chr. 4 - 41194313 - 41275321 on NCBI Build GRCm38 |
| Downstream | ubiquitin-associated protein 1 | Ubap1 |
2700092A01Rik, NAG20, UBAP-1, Ubap |
34.2kb | Anti-sense | Chr. 4 - 41348996 - 41389766 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Dcaf12
Genomic Map