Assay Details
| Gene Symbol: Slc28a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK085680.1 | - | 3 - 4 | 2026 | 88 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
RMI1, RecQ mediated genome instability 1, homolog (S. cerevisiae) |
Rmi1 |
4932432N11Rik, C79893 |
-142.2kb | Anti-sense | Chr. 13 - 58402248 - 58411149 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 4930455J16 gene | 4930455J16Rik | 152.8kb | Sense | Chr. 13 - 58741074 - 58762311 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Slc28a3
Genomic Map