Assay Details
| Gene Symbol: D930014O13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK086224.1 | - | 1 | 2350 | 75 | |
| AK086225.1 | - | 1 | 2351 | 75 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | empty spiracles homeobox 1 | Emx1 | -39.4kb | Anti-sense | Chr. 6 - 85187931 - 85204463 on NCBI Build GRCm38 | |
| Downstream | RAB11 family interacting protein 5 (class I) | Rab11fip5 |
9130206P09Rik, C75969, D6Ertd32e, GAF1, RIP11 |
88.3kb | Sense | Chr. 6 - 85334962 - 85374648 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

D930014O13Rik
Genomic Map