Assay Details
| Gene Symbol: Itpkc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK086754.1 | - | 1 | 2155 | 81 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 1191 | Mir1191 |
Mirn1191, mmu-mir-1191, mmu-mir-1191a |
-1.4kb | Sense | Chr. 7 - 27205536 - 27205655 on NCBI Build GRCm38 |
| Downstream | aarF domain containing kinase 4 | Adck4 | 0610012P18Rik | 18.6kb | Sense | Chr. 7 - 27228823 - 27257949 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Itpkc
Genomic Map