Assay Details
| Gene Symbol: Gm3414 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027993.1 | 1 | 456 | 91 | ||
| GenBank mRNA | AK088949.1 | - | 1 | 456 | 91 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 36401 |
Gm36401 | -8.7kb | Anti-sense | Chr. 5 - 45701997 - 45715716 on NCBI Build GRCm38 | |
| Downstream | RIKEN cDNA 4930405L22 gene | 4930405L22Rik | 199.2kb | Sense | Chr. 5 - 45926822 - 45932409 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gm3414
Genomic Map