Assay Details
| Gene Symbol: Gm4719 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045952.1 | 2 - 3 | 305 | 68 | ||
| GenBank mRNA | AK131717.1 | - | 2 - 3 | 306 | 68 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 4933400B14 gene | 4933400B14Rik | -53.3kb | Sense | Chr. 17 - 89320227 - 89327333 on NCBI Build GRCm38 | |
| Downstream | neurexin I | Nrxn1 |
1700062G21Rik, 9330127H16Rik, A230068P09Rik, mKIAA0578 |
651.4kb | Anti-sense | Chr. 17 - 90033644 - 91093100 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm4719
Genomic Map