Assay Details
| Gene Symbol: Dhh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK134149.1 | - | 1 - 2 | 121 | 89 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | Ras homolog enriched in brain like 1 | Rhebl1 |
1810036J22Rik, Rheb2 |
-11.9kb | Anti-sense | Chr. 15 - 98877760 - 98881497 on NCBI Build GRCm38 |
| Downstream | limb region 1 like | Lmbr1l |
1110013E13Rik, AA415653, D15Ertd735e, Limr |
8.7kb | Anti-sense | Chr. 15 - 98903914 - 98918550 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Dhh
Genomic Map