Assay Details
| Gene Symbol: Gm8541 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK136239.1 | - | 1 - 2 | 56 | 77 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
potassium voltage-gated channel, subfamily H (eag-related), member 7 |
Kcnh7 |
9330137I11Rik, Kv11.3, erg3 |
-382.8kb | Sense | Chr. 2 - 62699587 - 63184287 on NCBI Build GRCm38 |
| Downstream | fidgetin | Fign |
fi, fidget |
396.2kb | Sense | Chr. 2 - 63971508 - 64098038 on NCBI Build GRCm38 |
More Information
Set Membership: |
|
MicroArray Probes:
Agilent
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1988403 | AK136239.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 947333 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 591925 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 825316 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 15917 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 313567 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 880170 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 656321 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 658236 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 115069 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 717916 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 311623 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 165442 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 969806 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 27979 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 416018 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 169234 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 124380 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 112274 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 165128 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1988403 | AK136239.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04542493 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04542494 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04542525 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275784 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275881 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275852 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275653 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275752 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02208399 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275839 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275696 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275891 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01305918 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275740 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275591 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01543243 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275675 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275750 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275819 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275782 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275857 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275862 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275766 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275666 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275786 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275609 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275641 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275695 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02041833 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275703 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01830846 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275613 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275805 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275825 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275871 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275888 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275880 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275756 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275588 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02063483 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275851 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275815 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01271719 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275866 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275868 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275699 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275702 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275770 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275768 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275887 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275711 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275807 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275799 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275832 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275698 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275875 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275843 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275771 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275589 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275803 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275886 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275870 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275765 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275664 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275593 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275788 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275798 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275865 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275830 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275611 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275639 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275854 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275861 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275649 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275643 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275597 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275813 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275872 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275795 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275817 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275704 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275853 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01543238 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275763 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275844 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275603 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275762 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01682396 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275742 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01830852 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275656 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275829 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275809 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275882 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275595 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275672 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275790 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275654 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275604 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275745 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275772 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275846 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275658 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275748 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275615 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275760 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275743 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01373087 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275607 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01480686 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275797 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275837 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02041828 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275648 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275659 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02260613 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275811 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01543240 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275885 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275864 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275878 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275884 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275755 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275791 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275867 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275645 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275673 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275873 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275652 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275670 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275841 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275660 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275850 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275858 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21275890 | - | - |

Gm8541
Genomic Map