Assay Details
| Gene Symbol: Hbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK136633.1 | - | 1 | 553 | 89 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | component of oligomeric golgi complex 5 | Cog5 |
5430405C01Rik, C87247, D18362, GOLTC1, GTC90 |
Sense | Chr. 12 - 31654868 - 31937630 on NCBI Build GRCm38 | |
| Downstream |
protein kinase, cAMP dependent regulatory, type II beta |
Prkar2b |
AI451071, AW061005, PKARIIbeta, Pkarb2, RII(beta) |
29.7kb | Anti-sense | Chr. 12 - 31958450 - 32061343 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Hbp1
Genomic Map