Assay Details
| Gene Symbol: Park2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK137478.1 | - | 1 - 2 | 98 | 86 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | PARK2 co-regulated | Pacrg | 1700008H23Rik | -200.0 | Anti-sense | Chr. 17 - 10403012 - 10840191 on NCBI Build GRCm38 |
| Downstream | predicted gene 28505 | Gm28505 | 136.0kb | Anti-sense | Chr. 17 - 11076182 - 11090739 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Park2
Genomic Map