Assay Details
| Gene Symbol: Gm16325 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045949.1 | 3 - 4 | 835 | 80 | ||
| GenBank mRNA | AK139061.1 | - | 3 - 4 | 836 | 80 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | urate oxidase | Uox | AI663847 | -17.3kb | Sense | Chr. 3 - 146597149 - 146631483 on NCBI Build GRCm38 |
| Downstream |
protein kinase, cAMP dependent, catalytic, beta |
Prkacb | Pkacb | 77.9kb | Anti-sense | Chr. 3 - 146729579 - 146812946 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm16325
Genomic Map