Assay Details
| Gene Symbol: Gm10549 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045415.1 | 2 - 3 | 409 | 85 | ||
| GenBank mRNA | AK141051.1 | - | 2 - 3 | 410 | 85 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | neuronal regeneration related protein | Nrep |
AI325076, D0H4S114, Harp, P311, PTZ17, SEZ17 |
-6.7kb | Anti-sense | Chr. 18 - 33437019 - 33464029 on NCBI Build GRCm38 |
| Downstream | erythrocyte protein band 4.1 like 1 opposite strand | Epb4.1l4aos | 2410004N09Rik | 320.2kb | Sense | Chr. 18 - 33794892 - 33795989 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm10549
Genomic Map