Assay Details
| Gene Symbol: Rps18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK144398.1 | - | 37 | 88 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 5330416C01 gene | 5330416C01Rik | Gm532 | Anti-sense | Chr. 13 - 97712315 - 97778952 on NCBI Build GRCm38 | |
| Downstream |
predicted gene, 34388 |
Gm34388 | 114.2kb | Anti-sense | Chr. 13 - 97865811 - 97894001 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Rps18
Genomic Map