Assay Details
| Gene Symbol: Gm805 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_046081.1 | 3 - 4 | 509 | 74 | ||
| GenBank mRNA | AK144886.1 | - | 3 - 4 | 509 | 74 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | intraflagellar transport 43 | Ift43 |
1700019E19Rik, R75064 |
-7.9kb | Sense | Chr. 12 - 86082533 - 86162459 on NCBI Build GRCm38 |
| Downstream | G patch domain containing 2 like | Gpatch2l |
1700020O03Rik, 4732487G21Rik, A630090I05Rik, Kiaa1152 |
69.8kb | Sense | Chr. 12 - 86241838 - 86291793 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
This assay targets the same location as RTA transcript cluster(s) TC0600001203.rn
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2046045 | AK144886.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 18674 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 781703 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 27812 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 749574 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 252980 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 191644 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 144070 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 35240 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 424163 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 234325 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 306901 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 206906 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 59679 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 660183 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 137130 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 48365 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 171880 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P168118 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2046045 | AK144886.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P168118 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02569199 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02569168 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P168118 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28608624 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28608641 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28608622 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28608681 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28608675 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28608621 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28608635 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28608672 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28608528 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28608667 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28608552 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28608576 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28608671 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28608677 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28608555 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10345776 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28608678 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10196555 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28608688 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28608631 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28608632 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09846278 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28608669 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28608626 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28608612 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09958550 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09846269 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28608639 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28608545 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28608531 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10208031 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28608539 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28608674 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28608643 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10335842 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28608540 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28608642 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28608580 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28608526 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28608573 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28608542 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28608633 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09846270 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28608556 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28608616 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28608637 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28608581 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28608538 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28608685 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28608547 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28608550 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28608647 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10208221 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10335843 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28608670 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28608544 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10345767 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10227359 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10201645 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28608574 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28608646 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28608617 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28608689 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28608628 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28608537 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Clariom S Array | PSR1200006210.mm.1 | NR_046081.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200006210.mm.1 | NR_046081.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200006210.mm.1 | TC1200000847.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200006209.mm.1 | NR_046081.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200006209.mm.1 | NR_046081.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200006209.mm.1 | TC1200000847.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200006212.mm.1 | NR_046081.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200006212.mm.1 | NR_046081.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200006212.mm.1 | TC1200000847.mm.1 | MTA TC |

Gm805
Genomic Map