Assay Details
| Gene Symbol: Ube2d2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK146659.1 | - | 2 - 3 | 498 | 75 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | transmembrane protein 173 | Tmem173 |
2610307O08Rik, ERIS, MPYS, Mita, STING |
-52.9kb | Anti-sense | Chr. 18 - 35733678 - 35740554 on NCBI Build GRCm38 |
| Downstream | CXXC finger 5 | Cxxc5 | 4930415K17Rik | 26.1kb | Sense | Chr. 18 - 35828924 - 35861693 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Ube2d2a
Genomic Map