Assay Details
| Gene Symbol: Rbm46os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040381.1 | 3 - 4 | 355 | 96 | ||
| NR_040382.1 | 3 - 4 | 346 | 96 | |||
| GenBank mRNA | AK019784.1 | - | 2 - 3 | 306 | 96 | |
| AK161442.1 | - | 3 - 4 | 356 | 96 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | lecithin-retinol acyltransferase (phosphatidylcholine-retinol-O-acyltransferase) | Lrat |
1300010A18Rik, AI449251 |
-13.6kb | Anti-sense | Chr. 3 - 82892582 - 82903974 on NCBI Build GRCm38 |
| Downstream | fibrinogen gamma chain | Fgg |
3010002H13Rik, AI256424 |
66.8kb | Sense | Chr. 3 - 83007896 - 83015049 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2367235 | AK161442.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P122290 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2367235 | AK161442.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P193211 | AK161442.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P193211 | AK015378.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P343667 | AK019784.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P595794 | AK161442.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P595794 | AK015378.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P101814 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22282091 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22281776 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02491076 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22282083 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22282120 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22282108 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22282088 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22282113 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02308623 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02515188 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02515190 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22282095 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22281728 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22281729 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22282123 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22282117 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22281788 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22282092 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22282004 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02692392 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03009559 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02320436 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02308599 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22281726 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22281727 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02365291 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02308597 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22282112 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22281725 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02495293 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02945133 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22281781 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22281785 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22282087 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22282114 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02365286 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22282002 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22282098 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22282107 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22281789 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22282096 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22281999 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22282126 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22282102 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02645920 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03009563 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03009562 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22282109 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02320428 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT MG-430 PM Array Plate | 1431523_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1444784_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1431523_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1444784_at | - | - |
| Mouse Genome 430 2.0 Array | 1444784_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1431523_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0300004220.mm.1 | NR_040381.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300004220.mm.1 | NR_040381.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300004220.mm.1 | NR_040382.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300004220.mm.1 | NR_040382.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300004220.mm.1 | TC0300000639.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300004222.mm.1 | NR_040381.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300004222.mm.1 | NR_040381.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300004222.mm.1 | NR_040382.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300004222.mm.1 | NR_040382.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300004222.mm.1 | TC0300000639.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300004225.mm.1 | NR_040381.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300004225.mm.1 | NR_040381.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300004225.mm.1 | NR_040382.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300004225.mm.1 | NR_040382.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300004225.mm.1 | TC0300000639.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300004214.mm.1 | NR_040381.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300004214.mm.1 | NR_040381.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300004214.mm.1 | TC0300000639.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300004215.mm.1 | NR_040381.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300004215.mm.1 | NR_040381.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300004215.mm.1 | TC0300000639.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300004221.mm.1 | NR_040381.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300004221.mm.1 | NR_040381.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300004221.mm.1 | NR_040382.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300004221.mm.1 | NR_040382.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300004221.mm.1 | TC0300000639.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300004228.mm.1 | NR_040381.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300004228.mm.1 | NR_040381.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300004228.mm.1 | NR_040382.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300004228.mm.1 | NR_040382.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300004228.mm.1 | TC0300000639.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300004219.mm.1 | NR_040381.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300004219.mm.1 | NR_040381.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300004219.mm.1 | NR_040382.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300004219.mm.1 | NR_040382.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300004219.mm.1 | TC0300000639.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300004223.mm.1 | NR_040381.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300004223.mm.1 | NR_040381.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300004223.mm.1 | NR_040382.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300004223.mm.1 | NR_040382.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300004223.mm.1 | TC0300000639.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300004224.mm.1 | NR_040381.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300004224.mm.1 | NR_040381.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300004224.mm.1 | NR_040382.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300004224.mm.1 | NR_040382.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300004224.mm.1 | TC0300000639.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300004226.mm.1 | NR_040381.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300004226.mm.1 | NR_040381.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300004226.mm.1 | NR_040382.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300004226.mm.1 | NR_040382.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300004226.mm.1 | TC0300000639.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1431523_PM_at | - | - |
| Mouse Expression Set 430 | 1431523_at | - | - |
| Mouse Expression Set 430 | 1444784_at | - | - |
| Mouse Expression Set 430 | 1444784_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1431523_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1444784_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1431523_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1444784_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 391352 | - | - |

Rbm46os
Genomic Map