Assay Details
| Gene Symbol: Bnip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008238.3 | NP_001008239.1 | 10 | 5649 | 82 | |
| NM_016787.4 | NP_058067.2 | 11 | 5685 | 82 | ||
| GenBank mRNA | AK161900.1 | - | 1 | 3349 | 82 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA B230323A14 gene | B230323A14Rik | -178.0kb | Sense | Chr. 9 - 69761146 - 69830199 on NCBI Build GRCm38 | |
| Downstream |
general transcription factor II A, 2 |
Gtf2a2 |
TFIIA-gamma, TfIIg, Tfiia2 |
195.0 | Sense | Chr. 9 - 70011851 - 70022866 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Bnip2
Genomic Map