Assay Details
| Gene Symbol: Gm4221 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK163160.1 | - | 3 - 4 | 307 | 81 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | zinc finger protein 608 | Zfp608 |
4932417D18Rik, D430007A19Rik, Znf608 |
-54.8kb | Anti-sense | Chr. 18 - 54888045 - 54992598 on NCBI Build GRCm38 |
| Downstream | GRAM domain containing 3 | Gramd3 |
9030613F08Rik, 9130427A09Rik, AW061306, NS3TP2 |
1.3mb | Sense | Chr. 18 - 56387485 - 56503792 on NCBI Build GRCm38 |
More Information
Set Membership: |
|
MicroArray Probes:
Agilent
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 399753 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 362247 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 817340 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 919004 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 540834 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 141733 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 503522 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 542627 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 493767 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 909229 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 332284 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 394675 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 17301 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 842437 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 860626 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 52228 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 579453 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 727191 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 133138 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 489596 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 309041 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 750459 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 947168 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 97051 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 709502 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 184180 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 829491 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 458094 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 630952 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 408041 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 730661 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 505707 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 373407 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 675569 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 814853 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 943261 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 248009 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 55028 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 38475 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 89505 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 119407 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 12517 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2024313 | AK163160.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03480757 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09007304 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09007164 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09007176 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09007254 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09007196 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03480667 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03480726 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P09007254 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P09007196 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722406 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722423 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13853967 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722398 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722576 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722467 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722455 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722560 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722471 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722445 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13817739 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722386 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14078823 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722565 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722498 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722508 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722387 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722584 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13796672 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13817559 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13817740 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722519 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13817717 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722608 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722518 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722345 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722436 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722425 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722447 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722569 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722589 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722515 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722394 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722588 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722521 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722440 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722513 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14083597 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722441 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722385 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722346 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722547 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722514 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13729478 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13636708 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722570 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722438 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722499 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722449 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722503 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722501 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722340 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722591 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722490 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722525 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13617529 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722347 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722574 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722500 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722421 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722464 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722561 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722363 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722550 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722495 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722402 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14083599 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722489 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722545 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722582 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722396 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722359 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722389 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14127724 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722470 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722491 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722399 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722548 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13726628 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13733429 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722428 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722462 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722355 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722339 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722444 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13726610 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722361 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722453 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13645719 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722511 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722590 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722520 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722343 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722356 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722405 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722496 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722456 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722357 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722473 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722546 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722572 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722552 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13729474 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722465 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13726616 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722575 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722523 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13862883 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13607323 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722555 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722497 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31722558 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13853966 | - | - |

Gm4221
Genomic Map