Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 6307 | Gm6307 | EG622283 | -29.5kb | Anti-sense | Chr. 2 - 180385604 - 180401802 on NCBI Build GRCm38 |
| Downstream |
solute carrier organic anion transporter family, member 4a1 |
Slco4a1 |
OATP-E, Slc21a12 |
16.7kb | Anti-sense | Chr. 2 - 180456250 - 180474853 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Genomic Map