Assay Details
| Gene Symbol: Camkmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK172547.1 | - | 1 - 2 | 106 | 94 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | prolyl endopeptidase-like | Prepl |
2810457N15Rik, 9530014L06Rik, D030028O16Rik, mKIAA0436 |
-1.8kb | Anti-sense | Chr. 17 - 85062309 - 85090274 on NCBI Build GRCm38 |
| Downstream |
SIX homeobox 3, opposite strand 1 |
Six3os1 |
D17Mgi26, E130112H22Rik, Rncr1, Six3os |
505.3kb | Anti-sense | Chr. 17 - 85601932 - 85618396 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Camkmt
Genomic Map