Assay Details
| Gene Symbol: Fbxw11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK179454.1 | - | 1 - 2 | 54 | 71 | |
| AK179700.1 | - | 1 - 2 | 71 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | serine/threonine kinase 10 | Stk10 |
Gek1, Lok, mKIAA4026 |
-18.3kb | Sense | Chr. 11 - 32533266 - 32624595 on NCBI Build GRCm38 |
| Downstream | small integral membrane protein 23 | Smim23 | 1700008A04Rik | 167.5kb | Anti-sense | Chr. 11 - 32820376 - 32824594 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Fbxw11
Genomic Map