Assay Details
| Gene Symbol: Gm16973 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_046209.1 | 2 - 3 | 402 | 74 | ||
| NR_046210.1 | 2 - 3 | 334 | 74 | |||
| GenBank mRNA | AK045225.1 | - | 2 - 3 | 602 | 74 | |
| BC027359.1 | - | 1 - 2 | 208 | 74 | ||
| CO425816.1 | - | 2 - 3 | 402 | 74 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | M-phase phosphoprotein 8 | Mphosph8 |
1500035L22Rik, 4930548G07Rik, AU040635 |
Anti-sense | Chr. 14 - 56668240 - 56697430 on NCBI Build GRCm38 | |
| Downstream | paraspeckle protein 1 | Pspc1 |
5730470C09Rik, AI327109, AI449052, PSP1 |
23.1kb | Sense | Chr. 14 - 56722441 - 56778316 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm16973
Genomic Map