Assay Details
| Gene Symbol: Thrb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC042730.1 | - | 1 - 2 | 80 | 70 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
retinoic acid receptor, beta |
Rarb |
A830025K23, Hap, Nr1b2 |
-730.9kb | Anti-sense | Chr. 14 - 16430839 - 17082331 on NCBI Build GRCm38 |
| Downstream |
nuclear receptor subfamily 1, group D, member 2 |
Nr1d2 |
RVR, Rev-erb |
377.1kb | Anti-sense | Chr. 14 - 18204056 - 18239106 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Thrb
Genomic Map