Assay Details
| Gene Symbol: F630040K05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC043643.1 | - | 2 - 3 | 480 | 92 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 1600014C23 gene | 1600014C23Rik | -15.4kb | Anti-sense | Chr. 17 - 45731766 - 45733844 on NCBI Build GRCm38 | |
| Downstream |
predicted gene, 35692 |
Gm35692 | 29.8kb | Anti-sense | Chr. 17 - 45806192 - 45828882 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

F630040K05Rik
Genomic Map