Assay Details
| Gene Symbol: B230216N24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC049988.1 | - | 1 - 2 | 2557 | 61 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | gypsy retrotransposon integrase 1 | Gin1 |
4930429M06Rik, 8430411H10Rik, Gin-1, Zh2c2 |
-237.4kb | Anti-sense | Chr. 1 - 97770168 - 97793710 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 1810006J02 gene | 1810006J02Rik | 95.8kb | Anti-sense | Chr. 1 - 98131467 - 98144655 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

B230216N24Rik
Genomic Map