Assay Details
| Gene Symbol: BC051077 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC051077.1 | - | 1 - 2 | 289 | 68 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | BQ933808.1 | - | 288 | Chr. - - [-] on Build | 68 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | phosphatidylethanolamine binding protein 1 | Pebp1 |
HCNP, Pbp, Pbp1, Pbpr, Rkip |
-25.0kb | Sense | Chr. 5 - 117282651 - 117287564 on NCBI Build GRCm38 |
| Downstream | V-set and immunoglobulin domain containing 10 | Vsig10 | 3.1kb | Anti-sense | Chr. 5 - 117318596 - 117355006 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

BC051077
Genomic Map