Assay Details
| Gene Symbol: Klrb1-ps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_073569.1 | 4 - 5 | 104 | |||
| GenBank mRNA | DQ237930.1 | - | 4 - 5 | 414 | 104 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
C-type lectin domain family 2, member e |
Clec2e |
Clra, clr-a |
-22.1kb | Anti-sense | Chr. 6 - 129091816 - 129100903 on NCBI Build GRCm38 |
| Downstream |
C-type lectin domain family 2, member d |
Clec2d |
Clr-b, Clrb, Ocil |
51.2kb | Sense | Chr. 6 - 129180615 - 129186535 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Klrb1-ps1
Genomic Map