Assay Details
| Gene Symbol: Rarg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | M32073.1 | - | 1 - 2 | 378 | 57 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | integrin beta 7 | Itgb7 | Ly69 | -10.0kb | Sense | Chr. 15 - 102215994 - 102231958 on NCBI Build GRCm38 |
| Downstream | predicted gene 9918 | Gm9918 |
EG432988, ENSMUSG00000053724 |
26.9kb | Anti-sense | Chr. 15 - 102270518 - 102274128 on NCBI Build GRCm38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the M32073.1 transcripts.
Set Membership: |
|

Rarg
Genomic Map