Assay Details
| Gene Symbol: 2600006K01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK011165.1 | - | 1 - 2 | 227 | 52 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 219a-2 | Mir219a-2 |
Mir219-2, Mirn219-2, mir-219a-2, mmu-mir-219a-2 |
-23.0kb | Sense | Chr. 2 - 29845631 - 29845727 on NCBI Build GRCm38 |
| Downstream | outer dense fiber of sperm tails 2 | Odf2 |
AI848335, MMTEST29 |
19.9kb | Anti-sense | Chr. 2 - 29889720 - 29931746 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

2600006K01Rik
Genomic Map