Assay Details
| Gene Symbol: Rspo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK011587.2 | - | 1 | 850 | 64 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | angiopoietin 1 | Angpt1 |
1110046O21Rik, Ang-1, Ang1 |
-493.2kb | Anti-sense | Chr. 15 - 42424667 - 42676977 on NCBI Build GRCm38 |
| Downstream |
eukaryotic translation initiation factor 3, subunit E |
Eif3e |
48kDa, Eif3s6, Int6, eIF3-p46, eIF3-p48 |
78.8kb | Anti-sense | Chr. 15 - 43250040 - 43282736 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rspo2
Genomic Map