Assay Details
| Gene Symbol: 2610202C22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK011859.1 | - | 1 | 445 | 94 | |
| BM221021.2 | - | 1 | 94 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 9919 | Gm9919 |
ENSMUSG00000053739, Gm19459 |
-31.7kb | Anti-sense | Chr. 10 - 58239034 - 58252702 on NCBI Build GRCm38 |
| Downstream | LIM and senescent cell antigen-like domains 1 | Lims1 |
2310016J22Rik, 4921524A02Rik, AI507642, AU021743, AW551584, C430041B13Rik, Lims1l, PINCH, PINCH1 |
38.3kb | Sense | Chr. 10 - 58323435 - 58424691 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

2610202C22Rik
Genomic Map