Assay Details
| Gene Symbol: Dtna | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK012743.1 | - | 1 | 955 | 112 | |
| AK134543.1 | - | 1 | 1210 | 112 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | glyceraldehyde-3-phosphate dehydrogenase pseudogene | Gm7788 | EG665797 | -326.0kb | Sense | Chr. 18 - 23217596 - 23217818 on NCBI Build GRCm38 |
| Downstream | predicted gene 15972 | Gm15972 | OTTMUSG00000028454 | 205.6kb | Anti-sense | Chr. 18 - 23750447 - 23752305 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Dtna
Genomic Map