Assay Details
| Gene Symbol: Ror1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK012979.1 | - | 1 | 1350 | 89 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | phosphoglucomutase 2 | Pgm2 |
2610020G18Rik, AA407108, AI098105, Pgm-2, Pgm1 |
-455.1kb | Sense | Chr. 4 - 99929451 - 99987294 on NCBI Build GRCm38 |
| Downstream | ubiquitin-conjugating enzyme E2U (putative) | Ube2u | 34.4kb | Sense | Chr. 4 - 100478867 - 100550145 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ror1
Genomic Map