Assay Details
| Gene Symbol: Taf6l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK013112.1 | - | 1 | 353 | 197 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | interferon induced transmembrane protein 2 pseudogene | Gm2518 | D630022E15Rik | -3.6kb | Anti-sense | Chr. 19 - 8774485 - 8776962 on NCBI Build GRCm38 |
| Downstream | polymerase (RNA) II (DNA directed) polypeptide G | Polr2g |
2410046K11Rik, A230108L04Rik, C76415, RBP7, Rpo2-7l |
12.1kb | Sense | Chr. 19 - 8793129 - 8798557 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Taf6l
Genomic Map