Assay Details
| Gene Symbol: Meg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_003633.3 | 5 | 10651 | 102 | ||
| NR_027651.2 | 9 | 10639 | 102 | |||
| GenBank mRNA | AK013532.1 | - | 1 | 852 | 102 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 770 | Mir770 |
Mirn770, mir-770, mmu-mir-770 |
-3.2kb | Sense | Chr. 12 - 109563692 - 109563785 on NCBI Build GRCm38 |
| Downstream | microRNA 673 | Mir673 |
Mirn673, mir-673, mmu-mir-673 |
3.7kb | Sense | Chr. 12 - 109571990 - 109572080 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Meg3
Genomic Map