Assay Details
| Gene Symbol: 2900021C02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK013565.1 | - | 1 | 646 | 77 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ribonucleotide reductase M2 | Rrm2 |
AA407299, R2 |
-142.8kb | Sense | Chr. 12 - 24708254 - 24714146 on NCBI Build GRCm38 |
| Downstream | kinase D-interacting substrate 220 | Kidins220 |
3110039L19Rik, AI194387, AI316525, C330002I19Rik, mKIAA1250 |
117.0kb | Sense | Chr. 12 - 24974876 - 25063152 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

2900021C02Rik
Genomic Map