Assay Details
| Gene Symbol: Gpr85 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK013598.1 | - | 1 | 557 | 94 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA B630005N14 gene | B630005N14Rik | AI666701 | -156.5kb | Sense | Chr. 6 - 13625675 - 13677966 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 2610001J05 gene | 2610001J05Rik | C79299 | 33.8kb | Sense | Chr. 6 - 13869074 - 13871483 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gpr85
Genomic Map