Assay Details
| Gene Symbol: 2900046F13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK013655.1 | - | 1 | 953 | 71 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | DV045881.1 | - | 369 | Chr. - - [-] on Build | 71 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 10855 | Gm10855 | ENSMUSG00000075538 | -106.0kb | Sense | Chr. 2 - 6932547 - 6939595 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 1700061F12 gene | 1700061F12Rik | 2.1mb | Anti-sense | Chr. 2 - 9189515 - 9197484 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

2900046F13Rik
Genomic Map