Assay Details
| Gene Symbol: 3100003M19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK013928.1 | - | 1 | 1647 | 67 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | cadherin 2 | Cdh2 |
CDHN, N-cadherin, Ncad |
Sense | Chr. 18 - 16588877 - 16809049 on NCBI Build GRCm38 | |
| Downstream | predicted gene 15328 | Gm15328 |
D830029L11, OTTMUSG00000020196 |
216.9kb | Anti-sense | Chr. 18 - 16816407 - 16822783 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

3100003M19Rik
Genomic Map