Assay Details
| Gene Symbol: 3110021A11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_030776.1 | 1 | 646 | 65 | ||
| GenBank mRNA | AK014066.1 | - | 1 | 647 | 65 | |
| AK014105.1 | - | 1 | 629 | 65 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
wingless-type MMTV integration site family, member 5B |
Wnt5b |
AW545702, Wnt-5b |
-303.4kb | Anti-sense | Chr. 6 - 119432531 - 119544824 on NCBI Build GRCm38 |
| Downstream | RAD52 homolog (S. cerevisiae) | Rad52 | Rad52yh | 53.2kb | Sense | Chr. 6 - 119902207 - 119922823 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

3110021A11Rik
Genomic Map