Assay Details
| Gene Symbol: Cltc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK014236.1 | - | 1 | 655 | 109 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | peptidyl-tRNA hydrolase 2 | Ptrh2 |
A230072I16Rik, Bit1, CGI-147 |
-1.9kb | Anti-sense | Chr. 11 - 86683983 - 86692457 on NCBI Build GRCm38 |
| Downstream | DEAH (Asp-Glu-Ala-His) box polypeptide 40 | Dhx40 |
2410016C14Rik, ARG147, DDX40, PAD |
73.7kb | Sense | Chr. 11 - 86768849 - 86807660 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Cltc
Genomic Map