Assay Details
| Gene Symbol: 4632404M16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK014550.1 | - | 1 | 2748 | 62 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | nescient helix loop helix 2 | Nhlh2 |
6230401I09Rik, Hen2, NSCL2, Nscl-2, bHLHa34 |
-119.1kb | Sense | Chr. 3 - 102010110 - 102015492 on NCBI Build GRCm38 |
| Downstream |
vang-like 1 (van gogh, Drosophila) |
Vangl1 |
Kitenin, Lpp2, Stb2, Stbm |
18.4kb | Anti-sense | Chr. 3 - 102155899 - 102205011 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4632404M16Rik
Genomic Map