Assay Details
| Gene Symbol: 4632409D06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK014560.1 | - | 1 | 1956 | 85 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | zinc finger protein 326 | Zfp326 |
5730470H14Rik, ZAN75, Znf326 |
-42.9kb | Sense | Chr. 5 - 105876568 - 105915818 on NCBI Build GRCm38 |
| Downstream |
predicted gene, 32736 |
Gm32736 | 133.5kb | Sense | Chr. 5 - 106094943 - 106112121 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4632409D06Rik
Genomic Map